6-Bromo-4-chloro-8-fluoroquinoline 97%

Supplier: Ambeed
Warning

A553541-100MG A553541-1G A553541-250MG
77169-754EA 53.56 USD
77169-754 77169-756 77169-758
6-Bromo-4-chloro-8-fluoroquinoline 97%
6-Bromo-4-chloro-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrClFN
MW: 260.49 g/mol
MDL Number: MFCD09861346
CAS Number: 1019016-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR