3-Bromo-6-chloro-2-phenylimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A553193-100MG A553193-1G A553193-250MG
77197-522EA 142 USD
77197-522 77197-524 77197-526
3-Bromo-6-chloro-2-phenylimidazo[1,2-b]pyridazine 97%
3-Bromo-6-chloro-2-phenylimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₁₂H₇BrClN₃
MDL Number: MFCD09832527
CAS Number: 19601-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR