7-Chloro-6-fluoro-1H-indole 95%

Supplier: Ambeed
Warning

A553122-100MG A553122-250MG A553122-1G
77169-750EA 36 USD
77169-750 77169-752 77893-126
7-Chloro-6-fluoro-1H-indole 95%
7-Chloro-6-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MDL Number: MFCD12924666
CAS Number: 169673-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR