tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate 96%

Supplier: Ambeed
Warning

A553084-10G A553084-1G A553084-250MG A553084-5G A553084-25G
77169-742EA 254 USD
77169-742 77169-744 77169-746 77169-748 77594-686
tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate 96%
tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD17016202
CAS Number: 203661-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR