(S)-tert-Butyl 3-methoxypyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A552492-100MG A552492-1G A552492-250MG
77169-730EA 25 USD
77169-730 77169-732 77169-734
(S)-tert-Butyl 3-methoxypyrrolidine-1-carboxylate 95%
(S)-tert-Butyl 3-methoxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD16620895
CAS Number: 550371-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR