1-Aminopyridinium iodide 98%

Supplier: Ambeed
Warning

A552303-10G A552303-1G A552303-25G A552303-5G A552303-100G
77197-512EA 27 USD
77197-512 77197-514 77197-516 77197-518 77747-135
1-Aminopyridinium iodide 98%
1-Aminopyridinium iodide

Precaución: for Research Use Only

Formula: C₅H₇IN₂
MDL Number: MFCD00051893
CAS Number: 6295-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR