2-Amino-4,6-di-tert-butylphenol 98%
Supplier: Ambeed
|
Warning
|
A552142-100MG
A552142-250MG
A552142-5G
77169-726EA
66.95
USD
77169-726
77169-728
77684-396
2-Amino-4,6-di-tert-butylphenol 98%
2-Amino-4,6-di-tert-butylphenol
Precaución: For research use only.
Formula:
C₁₄H₂₃NO MW: 221.34 g/mol |
MDL Number:
MFCD00173890 CAS Number: 1643-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...