3-Chloro-2,6-difluoroaniline 98%

Supplier: Ambeed
Warning

A551259-1G A551259-250MG A551259-5G
77169-716EA 50.47 USD
77169-716 77169-718 77169-720
3-Chloro-2,6-difluoroaniline 98%
3-Chloro-2,6-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
MDL Number: MFCD06660173
CAS Number: 287172-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR