2,6-Dimethylthiophenol 97%

Supplier: Ambeed
Danger

A548998-1G A548998-25G A548998-5G
77197-484EA 6 USD
77197-484 77197-486 77197-488
2,6-Dimethylthiophenol 97%
2,6-Dimethylthiophenol

Precaución: for Research Use Only

Formula: C₈H₁₀S
MDL Number: MFCD00010021
CAS Number: 118-72-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR