7-Bromoquinolin-6-ol 95%

Supplier: Ambeed
Warning

A548948-100MG A548948-250MG A548948-1G
77169-710EA 90.64 USD
77169-710 77169-712 77613-730
7-Bromoquinolin-6-ol 95%
7-Bromoquinolin-6-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.05 g/mol
MDL Number: MFCD18413587
CAS Number: 84174-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR