α,α,α,2-Tetrafluoro-m-toluic acid 98%

Supplier: Ambeed
Warning

A548917-100G A548917-1G A548917-5G
77197-478EA 51 USD
77197-478 77197-480 77197-482
α,α,α,2-Tetrafluoro-m-toluic acid 98%
α,α,α,2-Tetrafluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MDL Number: MFCD00040980
CAS Number: 115029-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR