9-Oxabicyclo[6.1.0]-4-nonene 97%

Supplier: Ambeed
Warning

A547110-1G A547110-250MG A547110-5G
77197-466EA 46.35 USD
77197-466 77197-468 77197-470
9-Oxabicyclo[6.1.0]-4-nonene 97%
9-Oxabicyclo[6.1.0]-4-nonene

Precaución: for Research Use Only

Formula: C₈H₁₂O
MW: 124.18 g/mol
MDL Number: MFCD00086420
CAS Number: 637-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR