trans-Cinnamamide 98%

Supplier: Ambeed
Warning

A545839-100G A545839-25G
77197-460EA 34 USD
77197-460 77197-462 77197-464
trans-Cinnamamide 98%
trans-Cinnamamide

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00008033
CAS Number: 22031-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR