2-Methyl-6-methoxypyridine-3-boronic acid 97%

Supplier: Ambeed
Danger

Synonyms: (6-Methoxy-2-methylpyridin-3-yl)boronic acid, (6-methoxy-2-methyl-pyridin-3-yl)boronic Acid, 3-Borono-6-methoxy-2-methylpyridine, (6-methoxy-2-methylpyridin-3-yl)boranediol

A544187-100MG A544187-250MG A544187-1G
77169-684EA 46.35 USD
77169-684 77169-686 77684-387
2-Methyl-6-methoxypyridine-3-boronic acid 97%
2-Methyl-6-methoxypyridine-3-boronic acid

Precaución: For research use only.

Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
MDL Number: MFCD05662386
CAS Number: 459856-12-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR