4-Chloro-4'-ethynyl-1,1'-biphenyl 95%

Supplier: Ambeed
Warning

A544099-100MG A544099-1G A544099-250MG
77169-678EA 46.35 USD
77169-678 77169-680 77169-682
4-Chloro-4'-ethynyl-1,1'-biphenyl 95%
4-Chloro-4'-ethynyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₉Cl
MW: 212.67 g/mol
MDL Number: MFCD28099440
CAS Number: 57399-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR