5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isobenzofuran-1,3-dione 97%

Supplier: Ambeed
Warning

A543338-100MG A543338-1G A543338-250MG
77197-430EA 55.62 USD
77197-430 77197-432 77197-434
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isobenzofuran-1,3-dione 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isobenzofuran-1,3-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₅
MW: 274.07 g/mol
MDL Number: MFCD11858595
CAS Number: 849677-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR