1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Bis(palmitoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, L-DPPC, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, L-α-Lecithin, Dipalmitoyl, L-alpha-Dipalmitoyl phosphatidylcholine

A542590-1G A542590-250MG A542590-100MG
77197-414EA 121.54 USD
77197-414 77197-416 77617-308
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 95%
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

Precaución: for Research Use Only

Formula: C₄₀H₈₀NO₈P
MW: 734.05 g/mol
MDL Number: MFCD00036903
CAS Number: 63-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR