2-Oxa-6-azaspiro[3.4]octane hemioxalate 98%

Supplier: Ambeed
Warning

A542261-5G A542261-1G
77169-676EA 927.99 USD
77169-676 77893-104
2-Oxa-6-azaspiro[3.4]octane hemioxalate 98%
2-Oxa-6-azaspiro[3.4]octane hemioxalate

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₆
MDL Number: MFCD22422280
CAS Number: 1523570-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR