3-Amino-5-chloropyridine-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-chloropicolinamide

A541968-100MG A541968-250MG
77169-672EA 104.03 USD
77169-672 77169-674
3-Amino-5-chloropyridine-2-carboxamide 95%
3-Amino-5-chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD09037839
CAS Number: 27330-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR