Methyl 4-amino-2-chloro-5-methoxybenzoate 97%

Supplier: Ambeed
Warning

A541966-100MG A541966-1G A541966-250MG
77169-666EA 293.55 USD
77169-666 77169-668 77169-670
Methyl 4-amino-2-chloro-5-methoxybenzoate 97%
Methyl 4-amino-2-chloro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.63 g/mol
MDL Number: MFCD23707454
CAS Number: 1427390-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR