6-Methyl-2-phenyl-4,5-dihydropyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

A541918-1G A541918-5G
77169-662EA 16 USD
77169-662 77169-664
6-Methyl-2-phenyl-4,5-dihydropyridazin-3(2H)-one 98%
6-Methyl-2-phenyl-4,5-dihydropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD00117361
CAS Number: 4578-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR