6-Oxo-1-phenyl-1,6-dihydropyridine-3-carboxylic acid 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A541554-250MG
A541554-1G
77169-660EA
146.26
USD
77169-660
77684-384
6-Oxo-1-phenyl-1,6-dihydropyridine-3-carboxylic acid 98%, Ambeed.Inc
6-Oxo-1-phenyl-1,6-dihydropyridine-3-carboxylic acid
Precaución: For research use only.
Formula:
C₁₂H₉NO₃ MW: 215.20 g/mol |
MDL Number:
MFCD07369218 CAS Number: 77837-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...