2-Chloro-N-(cyclohexylcarbamoyl)acetamide 95%

Supplier: Ambeed
Warning

A541540-100G A541540-10G A541540-25G A541540-5G
77197-404EA 173.04 USD
77197-404 77197-406 77197-408 77197-410
2-Chloro-N-(cyclohexylcarbamoyl)acetamide 95%
2-Chloro-N-(cyclohexylcarbamoyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₂O₂
MW: 218.6800 g/mol
MDL Number: MFCD03976170
CAS Number: 16467-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR