4-Bromoindole-3-carboxylic Acid 98%

Supplier: Ambeed
Warning

A541205-1G A541205-100MG A541205-250MG A541205-5G
77169-658EA 56 USD
77169-658 77893-101 77893-102 77893-103
4-Bromoindole-3-carboxylic Acid 98%
4-Bromoindole-3-carboxylic Acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD09817565
CAS Number: 110811-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR