tert-Butyl 6-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%

Supplier: Ambeed
Warning

A541040-100MG A541040-50MG
77197-388EA 519.12 USD
77197-388 77197-390
tert-Butyl 6-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
tert-Butyl 6-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁FN₂O₃
MW: 320.36 g/mol
MDL Number: MFCD18433699
CAS Number: 1258638-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR