1,3-Bis(2,4,6-trimethylphenyl)imidazolium tetrafluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dimesityl-1H-imidazol-3-ium tetrafluoroborate

A539978-1G A539978-250MG A539978-5G A539978-25G
77197-380EA 28 USD
77197-380 77197-382 77197-384 77747-105
1,3-Bis(2,4,6-trimethylphenyl)imidazolium tetrafluoroborate 98%
1,3-Bis(2,4,6-trimethylphenyl)imidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₅BF₄N₂
MDL Number: MFCD04971186
CAS Number: 286014-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR