CID-755673 98%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-2,3,4,5-tetrahydro-1H-benzofuro[2,3-c]azepin-1-one, CID755673

A539848-10MG A539848-1MG A539848-50MG A539848-5MG
77169-648EA 46.35 USD
77169-648 77169-650 77169-652 77169-654
CID-755673 98%
CID-755673

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MW: 217.2206 g/mol
MDL Number: MFCD03828155
CAS Number: 521937-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR