(±)-1,2-Butanediol 99%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dihydroxybutane, (±)-1,2-Butylene glycol, (±)-1,2-Dihydroxy butane, 1,2-Butylene glycol, Butane-1,2-diol, 1,2-Butanediol

A539728-500G A539728-5G A539728-25G
77197-376EA 127.72 USD
77197-376 77197-378 77617-300
(±)-1,2-Butanediol 99%
(±)-1,2-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
MDL Number: MFCD00004570
CAS Number: 584-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR