2,4,6-Trimethylpyridinium-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: CPTS, 2,4,6-Trimethylpyridinium p-Toluenesulphonate, 2,4,6-Collidinium p-Toluenesulphonate

A538472-100G A538472-10G A538472-25G A538472-5G
77197-366EA 180.25 USD
77197-366 77197-368 77197-370 77197-372
2,4,6-Trimethylpyridinium-p-toluenesulfonate 98%
2,4,6-Trimethylpyridinium-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃S
MW: 293.38 g/mol
MDL Number: MFCD00012810
CAS Number: 59229-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR