6-Chloro-3-fluoro-2-iodopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Chlorol-3-fluoro-2-iodopyridine

A537762-5G A537762-1G
77169-640EA 753.99 USD
77169-640 77893-095
6-Chloro-3-fluoro-2-iodopyridine 95%
6-Chloro-3-fluoro-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClFIN
MDL Number: MFCD18257562
CAS Number: 1211590-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR