2-Phenylpyrimidin-5-amine 97%
Supplier: Ambeed
|
Warning
|
A537394-100MG
A537394-1G
A537394-250MG
77197-354EA
132
USD
77197-354
77197-356
77197-358
2-Phenylpyrimidin-5-amine 97%
2-Phenylpyrimidin-5-amine
Precaución: for Research Use Only
Formula:
C₁₀H₉N₃ MW: 171.19 g/mol |
MDL Number:
MFCD00520252 CAS Number: 59808-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...