Benzyl 4-methylbenzoate 95%

Supplier: Ambeed
Warning

A535959-10G A535959-5G
77169-634EA 71.07 USD
77169-634 77169-636
Benzyl 4-methylbenzoate 95%
Benzyl 4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
MDL Number: MFCD04347391
CAS Number: 5467-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR