1-(2-(Benzyloxy)ethyl)-4-(hydroxydiphenylmethyl)quinuclidin-1-ium bromide 95%
Supplier: Ambeed
|
Warning
|
A535937-10MG
A535937-1G
A535937-1MG
A535937-250MG
A535937-50MG
A535937-5MG
77169-622EA
19
USD
77169-622
77169-624
77169-626
77169-628
77169-630
77169-632
1-(2-(Benzyloxy)ethyl)-4-(hydroxydiphenylmethyl)quinuclidin-1-ium bromide 95%
1-(2-(Benzyloxy)ethyl)-4-(hydroxydiphenylmethyl)quinuclidin-1-ium bromide
Precaución: for Research Use Only
Formula:
C₂₉H₃₄BrNO₂ |
MDL Number:
MFCD27976798 CAS Number: 869113-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...