3,6-Dioxaoctamethylenediamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Bis-NH2-PEG2, 2,2'-(Ethylenedioxy)bis(ethylamine), 1,8-Diamino-3,6-dioxaoctane, 1,2-Bis(2-aminoethoxy)ethane
A535542-25G
A535542-5G
77197-352EA
11
USD
77197-352
77551-618
3,6-Dioxaoctamethylenediamine 98%
3,6-Dioxaoctamethylenediamine
Precaución: for Research Use Only
Formula:
C₆H₁₆N₂O₂ MW: 148.20 g/mol |
MDL Number:
MFCD00040474 CAS Number: 929-59-9 UN: 2735 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...