Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A534400-1G A534400-250MG A534400-5G
77169-616EA 123.6 USD
77169-616 77169-618 77169-620
Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate 98%
Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD17015873
CAS Number: 1095822-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR