4-(4-Nitrophenyl)piperidine 96%

Supplier: Ambeed
Warning

A533000-100MG A533000-1G A533000-250MG A533000-5G
77169-602EA 35.02 USD
77169-602 77169-604 77169-606 77169-608
4-(4-Nitrophenyl)piperidine 96%
4-(4-Nitrophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD06804549
CAS Number: 26905-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR