(Cyclopropylmethyl)methylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-(Cyclopropylmethyl)-N-methylamine Hydrochloride

A531513-100MG A531513-1G A531513-250MG A531513-5G
77169-592EA 33.99 USD
77169-592 77169-594 77169-596 77169-598
(Cyclopropylmethyl)methylamine hydrochloride 98%
(Cyclopropylmethyl)methylamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁N·ClH
MW: 121.608 g/mol
MDL Number: MFCD09759080
CAS Number: 77335-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR