(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine 96%

Supplier: Ambeed
Warning

A531097-100MG A531097-1G A531097-250MG
77197-330EA 33 USD
77197-330 77197-332 77197-334
(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine 96%
(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD06260669
CAS Number: 613656-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR