2-(((1-(tert-Butoxycarbonyl)piperidin-2-yl)methyl)(methyl)amino)acetic acid 95+%

Supplier: Ambeed
Warning

A530330-100MG A530330-1G A530330-250MG
77169-578EA 111.24 USD
77169-578 77169-580 77169-582
2-(((1-(tert-Butoxycarbonyl)piperidin-2-yl)methyl)(methyl)amino)acetic acid 95+%
2-(((1-(tert-Butoxycarbonyl)piperidin-2-yl)methyl)(methyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₂O₄
MW: 286.36 g/mol
MDL Number: MFCD21092975
CAS Number: 1353943-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR