Methyl 2-bromo-3,5-dichloroisonicotinate 97%

Supplier: Ambeed
Warning

A530218-100MG A530218-1G A530218-250MG
77197-324EA 85.49 USD
77197-324 77197-326 77197-328
Methyl 2-bromo-3,5-dichloroisonicotinate 97%
Methyl 2-bromo-3,5-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₄BrCl₂NO₂
MW: 284.92 g/mol
MDL Number: MFCD26406015
CAS Number: 1807028-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR