5-Oxohexanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Glurate, 5-Ketohexanoic Acid, 4-Acetylbutyric Acid

A529202-100G A529202-10G A529202-1G A529202-25G A529202-5G A529202-500G
77169-564EA 189 USD
77169-564 77169-566 77169-568 77169-570 77169-572 77893-075
5-Oxohexanoic acid 97%
5-Oxohexanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MDL Number: MFCD00004412
CAS Number: 3128-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR