3-(Aminomethyl)benzo[b]thiophene hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)-1-benzothiophene hydrochloride, (Benzo[b]thiophen-3-yl)methylamine hydrochloride, Benzo[b]thiophen-3-ylmethanamine hydrochloride

A528968-1G A528968-250MG A528968-5G
77197-314EA 67 USD
77197-314 77197-316 77197-318
3-(Aminomethyl)benzo[b]thiophene hydrochloride 95%
3-(Aminomethyl)benzo[b]thiophene hydrochloride

Precaución: for Research Use Only

Formula: C₉H₉NS·ClH
MDL Number: MFCD01566652
CAS Number: 55810-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR