4-Isoquinolineboronic acid neopentyl glycol ester 95%

Supplier: Ambeed
Warning

Synonyms: Isoquinoline-4-boronic acid, 2,2-dimethylpropane-1,3-diol cyclic ester, 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline

A528340-1G A528340-250MG A528340-5G
77169-558EA 87.99 USD
77169-558 77169-560 77169-562
4-Isoquinolineboronic acid neopentyl glycol ester 95%
4-Isoquinolineboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₆BNO₂
MDL Number: MFCD06659058
CAS Number: 844891-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR