KU55933 ≥96%

Supplier: Ambeed
Warning

A528135-10MG A528135-1MG A528135-5MG A528135-50MG
77197-308EA 44 USD
77197-308 77197-310 77197-312 77747-082
KU55933 ≥96%
KU55933

Precaución: for Research Use Only

Formula: C₂₁H₁₇NO₃S₂
MDL Number: MFCD08276985
CAS Number: 587871-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR