1-Pyridin-3-ylethylamine 97%

Supplier: Ambeed
Danger

Synonyms: 3-(1-Aminoethyl)pyridine, 1-(Pyridin-3-yl)ethanamine

A528075-1G A528075-5G
77169-554EA 29.87 USD
77169-554 77169-556
1-Pyridin-3-ylethylamine 97%
1-Pyridin-3-ylethylamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD01691641
CAS Number: 56129-55-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR