1-tert-Butyl 2-methyl 3-oxopyrrolidine-1,2-dicarboxylate 98%

Supplier: Ambeed
Warning

A527681-5G A527681-1G
77197-306EA 120 USD
77197-306 77572-680
1-tert-Butyl 2-methyl 3-oxopyrrolidine-1,2-dicarboxylate 98%
1-tert-Butyl 2-methyl 3-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MDL Number: MFCD24369719
CAS Number: 194924-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR