1-Hexyl-2,3-dimethylimidazolium tetrafluoroborate 98%
Supplier: Ambeed
|
Danger
|
A527506-25G
A527506-5G
77197-302EA
14
USD
77197-302
77197-304
1-Hexyl-2,3-dimethylimidazolium tetrafluoroborate 98%
1-Hexyl-2,3-dimethylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₁H₂₁BF₄N₂ |
MDL Number:
MFCD03427619 CAS Number: 384347-21-1 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...