1-(2-Bromoethoxy)-2-methylbenzene 95%

Supplier: Ambeed
Warning

A526809-1G A526809-250MG A526809-5G
77197-294EA 99.91 USD
77197-294 77197-296 77197-298
1-(2-Bromoethoxy)-2-methylbenzene 95%
1-(2-Bromoethoxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.08 g/mol
MDL Number: MFCD00082851
CAS Number: 18800-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR