4,4'-Dihydroxy-[1,1'-biphenyl]-3,3'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A526556-1G A526556-250MG A526556-5G A526556-25G
77169-548EA 15 USD
77169-548 77169-550 77169-552 77551-586
4,4'-Dihydroxy-[1,1'-biphenyl]-3,3'-dicarboxylic acid 97%
4,4'-Dihydroxy-[1,1'-biphenyl]-3,3'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₆
MDL Number: MFCD24432712
CAS Number: 13987-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR