1,6-Hexanediol dimethacrylate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A525882-100G A525882-25G A525882-500G
77197-288EA 21 USD
77197-288 77197-290 77197-292
1,6-Hexanediol dimethacrylate 98% stabilized
1,6-Hexanediol dimethacrylate

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₄
MDL Number: MFCD00015425
CAS Number: 6606-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR